Fluke® 50-Series II Thermocouple Thermometers
Rugged design with splash- and dust-resistant housing
  • Readout in °F, °C, or Kelvin (K)
  • Meters are ruggedly designed with a splash- and dust-resistant housing.
  • Sleep mode increases battery life
Dual backlit display provides readout of channel temperature, differential temperature (dual-input models), and minimum/maximum/average readings with time stamp.
ITEM(S) ON SALE
$377.99 - $602.99USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Data Logging Number Of Probe Channels Types Of Probes Accepted
Availability Pricing
Fluke 51-2 60HZ Thermocouple Thermometer with Single Input
Always in Stock
EW-91500-04 Mfr # FLUKE-51-2 60HZ
COMPARE
No 1 Type J, Type K, Type T
$377.99USD / Each
$419.99USD / Each
Fluke 52-2 60HZ Thermocouple Thermometer with Dual Inputs
Always in Stock
EW-91500-06 Mfr # FLUKE-52-2 60HZ
COMPARE
No 2 Type E, Type J, Type K, Type N, Type R, Type S, Type T
$476.99USD / Each
$529.99USD / Each
Fluke 53-2 B 60HZ Single-Input Datalogging Thermocouple Thermometer EW-91500-08 Mfr # FLUKE-53-2 B 60HZ
COMPARE
Yes 1 Type E, Type J, Type K, Type N, Type R, Type S, Type T
$521.99USD / Each
$579.99USD / Each
Fluke 54-2 B 60HZ Dual-Input Datalogging Thermocouple Thermometer
Always in Stock
EW-91500-10 Mfr # FLUKE-54-2 B 60HZ
COMPARE
Yes 2 Type E, Type J, Type K, Type N, Type R, Type S, Type T
$602.99USD / Each
$669.99USD / Each