This product is now discontinued, View Related Products
Fluke 718 Series Intrinsically Safe Pressure Calibrators
Pressure sourcea nd milliamp measurement capability to calibrate and maintain almost any pressure device with one tool
Integrated pump with media protection helps protect the pump from fluid contamination and allows pumps to be serviced in the field
Error % calculation for quicker pass / fail decision making in the field
Best in class mA accuracy (0.015%) for better measurement confidence and workload coverage
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Pressure Accuracy Pressure Resolution
Availability Pricing
Fluke 718EX 100 Intrinsically Safe Pressure Calibrators, -12 to 30 PSI EW-68450-91 Mfr # FLUKE-718EX 100
COMPARE
Call For Price
Fluke 718EX 30 Intrinsically Safe Pressure Calibrators, -12 to 200 PSI EW-68450-92 Mfr # FLUKE-718EX 30