Fluke 721EX Intrinsically Safe Precision Pressure Calibrator

Provide pressure calibration and temperature measurement for custody transfer applications
  • Both pressure inputs perform pressure switch tests
  • Large backlit graphic LCD displays up to three inputs simultaneously
  • Store five instrument setups for quick recall of routine tests
  • Choose between standard and intrinsically safe models
Dual-isolated pressure sensors make the Fluke 721 precision calibrator the ideal tool for gas custody transfer applications, allowing you to take simultaneous static and differential pressure measurements with one calibrator.

Intrinsically safe 721Ex models (16109-00 through -14) are ATEX Zone I and IECEx rated for use in hazardous environments.
$3,421.99USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Min Pressure Pressure Accuracy
Availability Pricing
Fluke 721EX-1603 Pressure Calibrator 16 PSIG to 300 PSIG EW-16109-01 Mfr # FLK-721EX-1603
COMPARE
16 psig sensor 1, 300 psig sensor 2 -14 psig sensor 1, -12 psig sensor 2 0.025% FS sensor 1, 0.025% FS sensor 2
$3,421.99USD / Each
Fluke 721EX-1630 Pressure Calibrator 16 PSIG to 3000 PSIG EW-16109-05 Mfr # FLK-721EX-1630
COMPARE
16 psig sensor 1, 3000 psig sensor 2 -14 psig sensor 1, 0 psig sensor 2 0.025% FS sensor 1, 0.025% FS sensor 2
$3,421.99USD / Each
Fluke 721EX-3615 Pressure Calibrator 36 PSIG to 1500 PSIG EW-16109-12 Mfr # FLK-721EX-3615
COMPARE
36 psig sensor 1, 1500 psig sensor 2 -14 psig sensor 1, 0 psig sensor 2 0.025% FS sensor 1, 0.025% FS sensor 2
$3,421.99USD / Each
Fluke 721EX-3630 Pressure Calibrator 36 PSIG to 3000 PSIG EW-16109-13 Mfr # FLK-721EX-3630
COMPARE
36 psig sensor 1, 3000 psig sensor 2 -14 psig sensor 1, 0 psig sensor 2 0.025% FS sensor 1, 0.025% FS sensor 2
$3,421.99USD / Each
Fluke 721EX-3650 Pressure Calibrator 36 PSIG to 5000 PSIG EW-16109-14 Mfr # FLK-721EX-3650
COMPARE
36 psig sensor 1, 5000 psig sensor 2 -14 psig sensor 1, 0 psig sensor 2 0.025% FS sensor 1, 0.035% FS sensor 2
$3,421.99USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat