Fluke 771, 772 and 773 Process mA Clamp Meters

Save time by not breaking the loop on 4-20 mA signal measurements
These innovative mA clamp meters are designed to measure, test and troubleshoot 4 to 20 mA control signals without breaking the mA loop. Make mA measurements in active control systems without disrupting process control. Save time and money troubleshooting process control loops.

All 771, 772, and 773 Fluke Models feature

  • Best in class 0.2 % accuracy

  • 0.01 mA resolution and sensitivity

  • Measure 4 to 20 mA signals without breaking the loop

  • Measure mA signals for PLC and control system analog I/O

  • Dual backlit display with both mA measurement and percent of 4 to 20 mA span

  • Measurement Spotlight illuminates hard to see wires in dark enclosures

  • Detachable clamp with extension cable for measurements in tight locations

  • Measure 10 to 50 mA signals in older control systems using the 99.9 mA range

  • Automatically change the 4 to 20 mA output for remote testing

  • Power saving features, auto-off, backlight timeout extend battery life

  • Hold function captures and displays changing measurements

  • Three year warranty for electronics; 1 year warranty for clamp cable assembly



Available for sale in the United States and Canada only.
ITEM(S) ON SALE
$939.38 - $1,830.99USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Measurement (mA) Max Measurement (VDC)
Availability Pricing
Fluke 772 Milliamp Process Clamp Meter with In-Circuit Measurement EW-20031-50 Mfr # FLUKE-772
COMPARE
24
$1,441.99USD / Each
Fluke 773 Milliamp Process Clamp Meter w/ VDC Measurement Capability EW-20031-52 Mfr # FLUKE-773
COMPARE
100 30
$1,830.99USD / Each
Fluke 771 Milliamp Process Clamp Meter with Case EW-23031-84 Mfr # FLUKE-771
COMPARE
100
$939.38USD / Each
$975.99USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat