This product is now discontinued, View Related Products
Fluke 975 AirMeter
Five powerful tools in one rugged, easy-to-use, handheld device
  • Store up to 2500 readings for later analysis
  • Calculates dewpoint, wet bulb temperature, % outside air, and air flow
Use this meter to optimize HVAC systems to minimize costs and increase occupant satisfaction and productivity. Unit will simultaneously display temperature, humidity, CO, and CO2. Datalogging feature will record 99 single-point readings or can be set to continuously monitor up to 25,000 records, leaving the technician free to perform other duties. A keypad lockout prevents unauthorized tampering.
Available for sale in the United States and Canada only.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Air Velocity (ft/min)
Availability Pricing
Fluke 975 Indoor Air Quality Meter EW-86570-10 Mfr # FLUKE-975
COMPARE
3000
Call For Price
Fluke 975V Indoor Air Quality Meter with Air Velocity EW-86570-12 Mfr # FLUKE-975V
COMPARE
3000