Fluke Calibration E-DWT-H Electronic Deadweight Testers
Get the precision and convenience of continuous real-time digital measurements!
  • Sets and reads any pressure value directly in any unit of measure, without moving weights
  • On-board AutoTest™ function lets you quickly define test points and adjust settings
  • Built-in filling and priming system
  • Interface with a PC or laptop for automated data acquisition
  • AutoRange function optimizes measurement and safety features
  • Low torque variable volume allows for pressure generation up to 30,000 psi with minimal physical effort
  • Compact and transportable—rechargeable battery pack for 8 hours of field operation
  • Switch Q-RPT ranges in seconds, eliminating piston-cylinder changes
  • No weights to load and unload; no minimum increment limited by smallest mass
  • Level, vibration, gravity, or temperature conditions will not affect readings
The E-DWT-H electronic deadweight tester generates and measures pressure for a variety of precision pressure calibration and testing applications. This system combines a high-performance electronic reference pressure monitor and manually operated pressure generation and control hardware in a single compact package. The electronic reference pressure monitor portion of the instrument can be controlled locally by the operator using the front panel display and keypad, or remotely via computer using ASCII character command strings over an RS-232 interface.
The 6531 models feature a single quartz reference pressure transducer (Q-RPT) that provides percent of reading performance from 10 to 100% of device full scale. The 6532 models feature a second Q-RPT that improves workload coverage with percent of reading performance from 1 to 100% of device full scale.
E-DWT-H testers comes with everything you need to perform high-level hydraulic testing right out of the box, including a vacuum fill kit to eliminate air pockets while filling and a test station adapter for mounting test devices without tape or wrenches.
Available for sale in the United States and Canada only
8 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Pressure Accuracy Pressure Resolution
Availability Pricing
Fluke Calibration 6531-7M Digital Deadweight Tester; 100 to 1000 psi EW-16106-50 Mfr # 6531-7M
COMPARE
1000 psi (7 MPa) Default: 0.01% of active range. User adjustable to 1 ppm of Q-RPT maximum or 10 ppm of active AutoRange, whichever is larger.
$28,490.00USD / Each
Fluke Calibration 6531-14M Digital Deadweight Tester; 200 to 2000 psi EW-16106-51 Mfr # 6531-14M
COMPARE
2000 psi (14 MPa) Default: 0.01% of active range. User adjustable to 1 ppm of Q-RPT maximum or 10 ppm of active AutoRange, whichever is larger.
$28,490.00USD / Each
Fluke Calibration 6531-20M Digital Deadweight Tester; 300 to 3000 psi EW-16106-52 Mfr # 6531-20M
COMPARE
3000 psi (20 MPa) Default: 0.01% of active range. User adjustable to 1 ppm of Q-RPT maximum or 10 ppm of active AutoRange, whichever is larger.
$28,490.00USD / Each
Fluke Calibration 6531-40M Digital Deadweight Tester; 600 to 6000 psi EW-16106-53 Mfr # 6531-40M
COMPARE
6000 psi (40 MPa) Default: 0.01% of active range. User adjustable to 1 ppm of Q-RPT maximum or 10 ppm of active AutoRange, whichever is larger.
$28,490.00USD / Each
Fluke Calibration 6531-70M Digital Deadweight Tester; 1000 to 10,000 psi EW-16106-54 Mfr # 6531-70M
COMPARE
10000 psi (70 MPa) Default: 0.01% of active range. User adjustable to 1 ppm of Q-RPT maximum or 10 ppm of active AutoRange, whichever is larger.
$28,490.00USD / Each
Fluke Calibration 6531-140M Digital Deadweight Tester; 2000 to 20,000 psi EW-16106-55 Mfr # 6531-140M
COMPARE
20000 psi (140 MPa) Default: 0.01% of active range. User adjustable to 1 ppm of Q-RPT maximum or 10 ppm of active AutoRange, whichever is larger.
$31,580.00USD / Each
Fluke Calibration 6532-70M Digital Deadweight Tester; 100 to 10,000 psi EW-16106-57 Mfr # 6532-70M
COMPARE
1000 psi (7 MPa) Default: 0.01% of active range. User adjustable to 1 ppm of Q-RPT maximum or 10 ppm of active AutoRange, whichever is larger.
$40,670.00USD / Each
Fluke Calibration 6532-140M Digital Deadweight Tester; 200 to 20,000 psi EW-16106-58 Mfr # 6532-140M
COMPARE
20000 psi (140 MPa) Default: 0.01% of active range. User adjustable to 1 ppm of Q-RPT maximum or 10 ppm of active AutoRange, whichever is larger.
$42,750.00USD / Each
Showing 1- 8 of 8