Fluke Calibration (formerly Hart Scientific) 1620A DewK High-Accuracy Thermohygrometers
Monitor and record conditions throughout your entire facility
  • Up to sixteen easily modified displays can be stored and recalled at the touch of a single button
  • Built-in RS-232 interface
Easily monitor and record conditions throughout your entire facility
The DewK meter accepts inputs from up to two sensors, which may be mounted directly on the unit or remotely (up to 100 feet away). All DewK sensors come with NVLAP accredited certificates of calibration for both temperature and humidity, complete with data and NIST traceability. Network as many DewKs as you want with the built-in Ethernet RJ45 jack, multiple units can be monitored from the same screen using the optional LogWare III client-server software. The Ethernet functionality also gives you the ability to remote connect over the internet, so you can monitor critical conditions while you’re away. The DewK is also available with wireless through an RF modem up to 100 ft away.
Available for sale in the United States and Canada only.
ITEM(S) ON SALE
$3,046.50 - $4,276.80USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Dew Point Range (° F) Max RH Range (%) Min RH Range (%)
Availability Pricing
Fluke Calibration 1620A-H Thermohygrometer, High Accuracy EW-37852-34 Mfr # 1620A-H-156
COMPARE
100 0
$3,477.60USD / Each
$3,864.00USD / Each
Fluke Calibration 1620A-S Thermohygrometer Kit , Standard EW-37852-35 Mfr # 1620A-S-156
COMPARE
100 0
$3,046.50USD / Each
$3,385.00USD / Each
Fluke Calibration 1621A-S Thermohygrometer Kit , Standard EW-37852-37 Mfr # 1621A-S-156
COMPARE
100 0
$4,276.80USD / Each
$4,752.00USD / Each