Fluke Calibration (formerly Hart Scientific) Chub High-Accuracy Lab Themometers
Display four channels with eight user-selected fields
  • Accuracy of ±0.002°F (±0.004°C) with RTD probe
  • Display four channels with eight user-selected data fields
  • Logs up to 8000 readings with time and date stamps on intervals from 0.1 sec-60 min
  • Reference-grade quiality and accuracy
Use this high-accuracy thermometer to read up to four different sensors simultaneously. Display up to eight user-selected data fields such as averages, standard deviations and spreads, probe type and serial number, T1-T2 functions, date, time and battery power levels. Save up to 10 screen configurations for easy recall. Line or battery powered, ideal for both benchtop and on-site readings. Push-button front panel brings up a simple menu that guides you through setup. Choose probe coefficients, sample intervals, communication settings, password protection, and more. RS-232 port allows data download to a computer or printer—order advanced software and printer below. What's included: battery, battery charger, basic software and cable
Available for sale in the United States and Canada only.
ITEM(S) ON SALE
$6,499.80USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
Fluke Calibration 1529 Precision Chub 4-Channel Thermometer; 2-PRT, 2 Thermocouple Probe EW-93202-00 Mfr # 1529-156
COMPARE
1900 -270 Precision Chub 4-Channel Thermometer; 2-PRT, 2 Thermocouple Probe
$6,499.80USD / Each
$7,222.00USD / Each
Fluke Calibration 1529-R Thermometer, 4 Thermistor/RTD Inputs EW-93202-10 Mfr # 1529-R-156
COMPARE
1900 -270 Thermometer, 4 Thermistor/RTD Inputs
$6,499.80USD / Each
$7,222.00USD / Each