Fluke Calibration (formerly Hart Scientific) Metrology Wells
Calibrate RTDs, PRTs, thermocouples, and thermistors with high-precision accuracy
  • Temperature range from -45 to 700°C
  • Optional ITS-90 reference input reads PRTs to ±0.006°C
These units combine bath-level performance with dry-well functionality and reference thermometry creating high precision metrology wells. The analog and digital control provides stability to ±0.005C, and dual zone uniformity control function (axial or radial) is as good as ±0.02°C over a 2.4” (60 mm) zone. The built-in input accepts 100, 25, and 10Ω PRTs, and ensures reading accuracies of thermometer probes from ±0.006°C at -200°C to ±0.027°C at 661°C (not including errors from the probe).
Units feature four different pre-programmed calibration tasks that allow up to eight temperature set points with ramp and soak times between each. The large LCD display, numeric keypad, and on-screen menus are easy to read and make programming simple. The display shows the block temperature, built-in reference thermometer temperature, cutout temperature, ramp rate, and stability criteria.
Available for sale in the United States and Canada only.
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy Temperature Resolution (° F)
Availability Pricing
Fluke Calibration 9171-A-156 Metrology Well, -30 to 155°C, 115 VAC EW-90919-06 Mfr # 9171-A-156
COMPARE
311
$13,393.00USD / Each
Fluke Calibration 9172-A-R-156 Metrology Well; 35 to 425 C/Built-In Ref EW-90919-12 Mfr # 9172-A-R-156
COMPARE
797
$12,252.00USD / Each