This product is now discontinued, View Related Products
Fluke Calibration (Hart Scientific) 5616 Series - Secondary reference PRT/RTD probes
5616 Series - Secondary reference PRT/RTD probes
High-purity platinum element and durability make these probes great for calibrating in the lab or in the field. When choosing a reference probe with a platinum element, there are two things you want to look at carefully: the short-term repeatability and the long-term drift. 5616s are among the best performing in their class with short-term repeatability better than ±0.010°C (±0.004°C is typical). In addition, the 5616 is specified to drift no more than ±0.010°C at the triple point of water when exposed up to its maximum temperature (420°C) for 100 hours. These specifications are given at k=3, and therefore
include a 99.8% confidence level.
Available for sale in the United States and Canada only.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Connector Type Max Temperature (° F) Min Temperature (° F)
Availability Pricing
Fluke Calibration 5616-12-L High Accuracy Temperature Probe Secondary PRT 100 O EW-90448-94 Mfr # 561612-L
COMPARE
Mini Spade Lug 788 -328
Call For Price
Fluke Calibration 3277531 PRT/RTD Probe with NIST-Traceable Certificate, 0.25" Dx12" L EW-90450-38 Mfr # 5616-12-A
COMPARE
Info-Con 788 -328
Call For Price
Fluke Calibration 3032648 PRT/RTD Bath Probe with NIST-Traceable Certificate, 0.25" Dx12" L; Wire EW-90450-39 Mfr # 5616-12-B
COMPARE
Stripped Wire 788 -328
Call For Price
Fluke Calibration 3032653 PRT/RTD Bath Probe with NIST-Traceable Certificate, 0.25" Dx12" L; 5-Pin EW-90450-40 Mfr # 5616-12-D
COMPARE
5-Pin DIN 788 -328
Call For Price
Fluke Calibration 3032675 PRT/RTD Probe with NIST-Traceable Certificate, 0.25" Dx12" L EW-90450-41 Mfr # 5616-12-I
COMPARE
Info-Con 788 -328
Call For Price
Fluke Calibration 3376961 PRT/RTD Bath Probe, 0.25" Dia x 12" L; I-Con EW-90450-42 Mfr # 5616-12-P
COMPARE
Info-Con 788 -328