Fluke Calibration (Hart Scientific) 5622 Series - Fast Response PRT/RTD probes
5622 Series - Fast response PRT/RTD probes
Designed for special temperature measurement applications requiring fast response or short immersion over a wide temperature range. These high-quality wire-wound sensors are small allowing heat to transfer to the sensors quickly, with a time constants as fast as 0.4 seconds. Immersion requirements for these probes is also a plus, ranging from just 10 mm to 64 mm (0.4 to 2.5 inch), depending on the model.
Available for sale in the United States and Canada only.
ITEM(S) ON SALE
$666.90USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Description
Availability Pricing
Fluke Calibration 3377019 Fast Response PRT Probe, 10" Info-Con EW-90450-61 Mfr # 5622-10-P
COMPARE
Info-Con 662 Fast Response PRT Probe, 10\" Info-Con
$666.90USD / Each
$741.00USD / Each