Fluke Calibration (Hart Scientific) 5626 Series - Secondary standard PRT/RTD probes
5626 Series - Secondary standard PRT/RTD probes: These probes fill the gap between affordable, but temperature-limited secondary PRTs and more expensive, highly accurate SPRTs. Using a regular PRT at temperatures above 500 °C exposes the platinum to contamination. If the PRT is used as a reference or calibration standard, contamination is a major problem. SPRTs, which are more expensive and delicate, can handle the higher temperatures, but with greater risk to the instrument due to shock, contamination, or mishandling. The 5626 is designed to reduce the contamination risk through the use of internal protection while not impairing performance The 5626 is nominally 100W.
Available for sale in the United States and Canada only.
$2,819.00 - $3,570.00USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
Fluke Calibration 5626-12-D PRT Probe; 0.25x12" L EW-90449-45 Mfr # 5626-12-D
COMPARE
1222
$2,819.00USD / Each
Fluke Calibration 5626-12-L PRT Probe; 0.25x12" L EW-90449-47 Mfr # 5626-12-L
COMPARE
Mini Spade Lug 1228
$2,819.00USD / Each
Fluke Calibration 1653830 PRT/RTD Bath Probe, 0.25" Dia x 12" L; Wire EW-90450-66 Mfr # 5626-12-B
COMPARE
Stripped Wire 1228
$2,819.00USD / Each
Fluke Calibration 1653875 PRT/RTD Bath Probe, 0.25" Dia x 12" L EW-90450-67 Mfr # 5626-12-I
COMPARE
Info-Con 1228
$2,819.00USD / Each
Fluke Calibration 3377055 PRT/RTD Bath Probe, 0.25" Dia x 12" L EW-90450-68 Mfr # 5626-12-P
COMPARE
Info-Con 1228
$2,819.00USD / Each
Fluke Calibration 1653918 PRT/RTD Bath Probe, 0.25" Dia x 12" L; Spade EW-90450-69 Mfr # 5626-12-S
COMPARE
Spade Lugs 1228
$2,819.00USD / Each
Fluke Calibration 1653929 PRT/RTD Bath Probe, 0.25" Dia x 15" L; Wire EW-90450-70 Mfr # 5626-15-B
COMPARE
Stripped Wire 1228
$2,888.00USD / Each
Fluke Calibration 1653934 PRT/RTD Bath Probe, 0.25" Dia x 15" L; 5-Pin EW-90450-71 Mfr # 5626-15-D
COMPARE
5-Pin DIN 1228
$2,888.00USD / Each
Fluke Calibration 1653965 PRT/RTD Bath Probe, 0.25" Dia x 15" L EW-90450-72 Mfr # 5626-15-I
COMPARE
Info-Con 1228
$2,888.00USD / Each
Fluke Calibration 1653983 PRT/RTD Bath Probe, 0.25" D x 15" L; Mini-Lug EW-90450-73 Mfr # 5626-15-L
COMPARE
Mini Spade Lug 1228
$2,888.00USD / Each
Fluke Calibration 3377062 PRT/RTD Bath Probe, 0.25" Dia x 15" L EW-90450-74 Mfr # 5626-15-P
COMPARE
Info-Con 1228
$2,888.00USD / Each
Fluke Calibration 1654000 PRT/RTD Bath Probe, 0.25" Dia x 15" L; Spade EW-90450-75 Mfr # 5626-15-S
COMPARE
Spade Lugs 1228
$2,888.00USD / Each
Fluke Calibration 2451974 PRT/RTD Bath Probe, 0.25" Dia x 20" L; Wire EW-90450-76 Mfr # 5626-20-B
COMPARE
Stripped Wire 1228
$3,570.00USD / Each
Fluke Calibration 2451988 PRT/RTD Bath Probe, 0.25" Dia x 20" L; 5-Pin EW-90450-77 Mfr # 5626-20-D
COMPARE
5-Pin DIN 1228
$3,570.00USD / Each
Fluke Calibration 2452015 PRT/RTD Bath Probe, 0.25" Dia x 20" L EW-90450-78 Mfr # 5626-20-I
COMPARE
Info-Con 1228
$3,570.00USD / Each
Fluke Calibration 2452032 PRT/RTD Bath Probe, 0.25" D x 20" L; Mini-Lug EW-90450-79 Mfr # 5626-20-L
COMPARE
Mini Spade Lug 1228
$3,570.00USD / Each
Fluke Calibration 3377070 PRT/RTD Bath Probe, 0.25" Dia x 20" L EW-90450-80 Mfr # 5626-20-P
COMPARE
Info-Con 1228
$3,570.00USD / Each
Fluke Calibration 2452059 PRT/RTD Bath Probe, 0.25" Dia x 20" L; Spade EW-90450-81 Mfr # 5626-20-S
COMPARE
Spade Lugs 1228
$3,570.00USD / Each
Showing 1- 18 of 18