Fluke Calibration TQSoft and TQAero Thermal Validation Software
Compliance made easy
This calibration and validation software simplifies test setup with step-by-step guides and dropdown lists of available options. Automatic and manual sensor calibrations for multichannel temperature measurement systems determine mulitple point offsets for each channel. The software assists with equipment setup, such as data logger channel configuration and temperature source communications. Validation test development options include 3-D chamber space diagrams with multicolor temperature gradients based on data from multiple sensors and real-time temperature displays. Once complete, test data can be played back in graph form showing the change in the temperature map over time.
Sample tests are included for practice and to use as templates in devising your own tests. A large library of reporting templates make report generation quick and easy. From cover pages to calibration certificates, the templates can be merged into a full report and saved or printed. All authorized changes, from equipment preparation and sensor calibration to final reports, are documented with full details of the changes and any actions performed providing comprehensive data security.
Choose TQSoft for pharmaceutical process analysis and validation, or TQAero for aerospace furnace validation.
For sale in US and Canada only
ITEM(S) ON SALE
$869.40 - $3,065.40USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Fluke Calibration TQSoft Thermal Validation Software with IQ/OQ Documents EW-16114-00 Mfr # TQSOFT-IQ/OQ
COMPARE
Thermal Validation Software with IQ/OQ Documents
$3,065.40USD / Each
$3,406.00USD / Each
Fluke Calibration TQAero  Thermal Validation Software EW-16114-01 Mfr # TQAERO
COMPARE
Thermal Validation Software
$2,024.10USD / Each
$2,249.00USD / Each
Fluke Calibration Thermal Validation Reference Binder on CD EW-16114-99 Mfr # VAL-BNDR-TQS
COMPARE
Thermal Validation Reference Binder on CD
$869.40USD / Each
$966.00USD / Each