Fluke CV-Series ClirVu Thermal Inspection Windows
Increase the safety of your personnel and reduce the time and cost of preventive maintenance
  • Quick and easy install – a standard hole punch and a torque wrench is all you need
  • Easy flip-open hatch for faster inspections
  • Simple thumb screw releases the permanently-hinged cover
  • Help reduce the vast majority of arc flash occurrences
Superior arc blast protection:
– High temperature silicon gaskets
– Mounting and door latches provide high pressure gasket compression
– Die cast components are manufactured with the highest strength alloys (ZA-27 and EZAC)
– Jam nut screws are made from 4037 steel, heat treated to 160,000 psi tensile
• Torture Tested to the highest test ratings
– IEEE C37.20.7 63 kA Arc tested at KEMA, UL
50/50E/50V, UL1558, IEC60529-1: IP67, IEC
60068, NEMA 4/12, CSA C22.2 NO. 14-13:2012, and CE
• Quick and easy installation
– Installation takes 5 minutes with one technician
– Only one hole necessary with standard Greenlee punch.
– Rear access required for punch connection only (Easily accomplished with one technician)
– Installs from front; no template needed; no rear access required
– Grounds instantly to metal enclosure with uniqe AutoGround process
– Sharp teeth on jam nuts assure four gas-tight ground connections
– All components have electrically conductive finishes for optimum grounding
• Quarter turn latch easily opens hinged cover
• Corrosion and UV resistance for challenging outdoor environments:
– All hardware is made from stainless steel or zinc plated steel
– Die cast components are plated with black chromium over phosphorous Electroless nickel
ITEM(S) ON SALE
$225.00 - $450.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Detector Size Max Temperature (° F) Pixels / Thermal Resolution
Availability Pricing
Fluke CV200 CirVu 2" Infrared Inspection Window EW-39750-61 Mfr # FLUKE-CV200
COMPARE
$225.00USD / Each
$250.00USD / Each
Fluke CV300 CirVu 3" Infrared Inspection Window EW-39750-63 Mfr # FLUKE-CV300
COMPARE
$315.00USD / Each
$350.00USD / Each
Fluke CV301 CirVu IR Inspection Window; 3" Dia with Security La Thermocouple Probe h EW-39750-64 Mfr # FLUKE-CV301
COMPARE
$315.00USD / Each
$350.00USD / Each
Fluke CV400 CirVu 4" Infrared Inspection Window EW-39750-65 Mfr # FLUKE-CV400
COMPARE
$450.00USD / Each
$500.00USD / Each
Fluke CV401 CirVu IR Inspection Window; 4" Dia with Security La Thermocouple Probe h EW-39750-66 Mfr # FLUKE-CV401
COMPARE
$450.00USD / Each
$500.00USD / Each