Fluke ii900 Industrial Sonic Imager
Turn sound waves into real-time images to locate leaks with ease
  • Quickly detect leaks in noisy or hard to reach environments
  • Easily view leaks on a large 7” LCD touch screen with overlaid SoundMap™
  • Reduce leak detection time and energy costs by accurately scanning large areas
  • Rugged IP40 unit rating with IP51 sensor head rating — ideal for industrial applications
Effectively detect and pinpoint leaks of compressed air, vacuum, steam, or other gases with the Fluke ii900 Sonic Industrial Imager. This leak detector imager uses an array of ultrasonic microphones in combination with SoundMap™ technology blended over real images to provide an image of the leak.
The Fluke ii900 imager isolates the frequency of leaks or vacuums, allowing detection in noisy production environments or from a distance. With a large array of microphones, the imager can quickly scan a large area. The ultrasonic detection range allows for use indoors and outdoors as measurements are not dependent on ambient or gas temperatures.
$23,349.99 - $25,427.99USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Decibel (Db) High Range (dB) Decibel (Db) Low Range (dB) Description
Availability Pricing
Fluke ii900 Sonic Industrial Imager with 1 Yr Premium Care EW-20051-25 Mfr # FLUKE-II900/FPC
COMPARE
120 16 Sonic Industrial Imager with 1 Yr Premium Care
$25,427.99USD / Each
Fluke ii900 Sonic Industrial Imager EW-39749-41 Mfr # FLK-ii900
COMPARE
120 16 Fluke ii900 Sonic Industrial Imager
$23,349.99USD / Each