Fluke® MDA-550 Series III Motor Drive Analyzers
Eliminate the hassle of setting up complex motor-drive troubleshooting measurements
  • Ensure critical data is captured automatically with preset measurement profiles
  • Reduce setup complexity with graphical step-by-step connection diagrams and guided testing
  • Quickly create reports to document troubleshooting and collaborative work with built-in report writing capabilities
  • Three powerful tools in one—motor-drive analyzer, waveform analyzer, and recording data logger
  • Safety rating of CAT IV-600 V/CAT III-1000 V
The Fluke® MDA-550 Series III motor drive analyzer saves time and eliminates the hassle of setting up complex measurements, while simplifying motor-drive troubleshooting. This analyzer is engineered to help you quickly and easily test and troubleshoot typical problems on three-phase and single-phase inverter type motor-drive systems.
Measure key motor-drive parameters including voltage, current, DC Bus voltage level and AC ripple, voltage and current unbalance and harmonics, voltage modulation, and motor shaft voltage discharges.
The on-screen information and setup guidance make it easy to configure the analyzer and get the drive measurements you need to make better maintenance decisions. Simply select a test and the step-by-step guided measurements show you where to make voltage and current connections, while the preset measurement profiles ensure you will capture all the data you need for each critical motor-drive section. Built-in report generator quickly and easily generated as-found and as-left reports with confidence.
$9,388.99 - $10,213.99USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Channels Frequency Range (Function Generator) Description
Availability Pricing
Fluke MDA-550-III Motor Drive Analyzer, 4 Channels, 500 MHz EW-20041-84 Mfr # FLUKE-MDA-550-III
COMPARE
4 1.000 Hz to 500 MHz MDA-550-III Motor Drive Analyzer, 4 Channels, 500 MHz
$9,388.99USD / Each
Fluke MDA-550-III Motor Drive Analyzer with 1 Yr Premium Care, 4 Channels, 500 MHz EW-20051-28 Mfr # FLUKE-MDA-550/FPC
COMPARE
4 1.000 Hz to 500 MHz MDA-550-III Motor Drive Analyzer with 1 Yr Premium Care, 4 Channels, 500 MHz
$10,213.99USD / Each