This product is now discontinued, View Related Products
Fluke TiS-Series Entry Level Performance Thermal Imagers
Built for performance with features that help you easily and quickly identify problems
  • Built for performance with features that help you easily and quickly identify problems
The TiS Performance Series thermal imagers are fully radiometric. They measure and store temperatures at every point in the image. IR-Fusion® technology provides enhanced problem detection and analysis capabilities through the blending of visual and infrared images. The built-in laser pointer also helps associate the physical target with the hot spot on the thermal image, indicating where a problem exists. Designed to withstand a 6.5-ft (2-m) drop.
These imagers come standard with a Wi-Fi connection. To activate the Wi-Fi function, simply download the Fluke Viewer app to your Apple® mobile device and link to your thermal imager. Then transfer images to your mobile device for further analysis, creating reports, and e-mailing results. A Bluetooth® connection is also standard on the TiS40 model and above. You can link the camera to your existing Bluetooth® headset (not included), allowing for record notes to be added to an image.
Fast and convenient menu navigation using either rugged touch screen or optimized tactile buttons. All data is stored in the 4 GB internal memory or on a removable 4 GB micro SD memory card. Additional imager features include large built-in camera lens cover and SmartView® software. SmartView software is a powerful modular suite of software tools for viewing, annotating, editing, and analyzing infrared images.
Fluke Connect® with ShareLive video™—make the best decisions faster than ever before by viewing all temperature, mechanical, electrical and vibration measurements for each equipment asset in one place. Get started saving time and increasing your productivity. Fluke Connect with ShareLive video call is the only wireless measurement system that lets you stay in contact with your entire team without leaving the field. The Fluke Connect mobile app is available for Android™ and iOS and works with over 20 different Fluke products.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Detector Size Max Temperature (° F) Pixels / Thermal Resolution
Availability Pricing
Fluke TiS Performance Series Thermal Imager (80 x 60) EW-39749-29 Mfr # FLK-TIS10 9HZ
COMPARE
80 x 60 662 4,800
Call For Price
Fluke TiS Performance Series Thermal Imager (120 x 90) EW-39749-30 Mfr # FLK-TIS20 9HZ
COMPARE
120 x 90 662 10,800
Call For Price
Fluke TiS Performance Series Thermal Imager (160 x 120) EW-39749-31 Mfr # FLK-TIS40 9HZ
COMPARE
160 x 120 662 19,200
Call For Price
Fluke TiS Performance Series Thermal Imager (160 x 120) with Manual Focus EW-39749-32 Mfr # FLK-TIS45 30HZ
COMPARE
160 x 120 662 19,200