FMI Compact Metering Pumps
Compact design RH pump with synchronous motor assembly
  • Drift-free performance independent of load variations or fluctuations in line voltage
  • Micrometer-like fine adjustment using an easy grip flow control ring graduated in 450 divisions
  • Choice of 150, 300, and 600 RPM through a simple and safe belt arrangement change
Forward-Off-Reverse switch for instant flow direction control
$935.75 - $998.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Flow Rate (Strokes/min) Power (VAC)
Availability Pricing
FMI Low-Flow Compact Metering Pump, 0 to 0.05 mL/revolution; 115 VAC, 60 Hz EW-07115-10 Mfr # RHSY0CKC
COMPARE
30 0.05 115
$935.75USD / Each
FMI Low-Flow Compact Metering Pump, 0 to 0.1 mL/revolution; 115 VAC, 60 Hz EW-07115-25 Mfr # RHSY1CKC
COMPARE
60 0.1 115
$998.00USD / Each
FMI Low-Flow Compact Metering Pump, 0 to 0.1 mL/revolution; 230 VAC, 50/60 Hz EW-07115-27 Mfr # RHSY1CKC2
COMPARE
60 0.1 230
$980.50USD / Each