FMI Single-Head, Fixed-Speed AC Drives
No valves to clog or wear out
Series Q drives are interchangeable with Series Q pump heads. These fixed-speed AC drives feature a TEFC motor. Pedestal drive model 07104-40 is for chain-, belt-, or shaft-coupling to rotating machine elements or to your motor. Build your own pump systems using interchangeable heads, drives, and accessories. Pump heads sold separately.
$353.10 - $1,157.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Motor RPM (rpm) Motor hp Description
Availability Pricing
COMPARE
1725 1/25 Series Q Drive, Single Head, Fixed Speed, TEFC, 1725 rpm; 110 VAC, 60 Hz
$1,082.50USD / Each
COMPARE
1725 1/25 Series Q Drive, Single Head, Fixed Speed, TEFC, 1725 rpm; 220 VAC, 50/60 Hz
$1,157.00USD / Each
COMPARE
1800 Requires motor with 20 oz-in of torque Series Q Drive, Single Head, Fixed Speed, Pedestal, 1800 rpm
$353.10USD / Each