FMI Valveless Piston Pump Heads, Series Q
No valves to clog or wear out
  • Build your own pump systems using interchangeable heads, drives, and accessories
  • Pump fluids with viscosities up to 500 cp
  • Low dead volume
These FMI Series Q valveless piston pump heads give you options to handle a wide range of fluids at pressures up to 100 psi. The interchangeable pump head modules provide numerous flow and material combinations for optimal versatility. Choose from a combination of ceramic, PVDF, carbon, and stainless steel wetted materials for an inert and corrosion resistant fluid path, ideal for your specific application. These materials offer broad fluid compatibility for dependable long-life operation.
$433.80 - $784.75USD / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Flow Rate (Strokes/min) Max Pressure (PSI)
Availability Pricing
FMI Series Q Valveless Piston Pump Head, Stainless Steel, Stainless Steel, Carbon; 0.08 mL/rev EW-07104-50 Mfr # FMI202 / Q0SSY
COMPARE
48 600 100
$634.00USD / Each
FMI Series Q Valveless Piston Pump Head, Stainless Steel, PVDF, Carbon; 0.08 mL/rev EW-07104-52 Mfr # FMI316 / Q0SKY
COMPARE
48 600 60
$476.50USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, PVDF, Ceramic; 1.28 mL/rev EW-07104-53 Mfr # FMI217 / Q3CKC
COMPARE
768 600 60
$601.25USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, PVDF, Ceramic; 0.32 mL/rev EW-07104-54 Mfr # FMI352 / Q1CKC + (2) R412-2K
COMPARE
192 600 60
$483.60USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, Stainless Steel, Ceramic; 0.32 mL/rev EW-07104-56 Mfr # FMI205 / Q1CSC
COMPARE
192 600 100
$595.00USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, PVDF, Carbon; 0.32 mL/rev EW-07104-58 Mfr # FMI358 / Q1CKY
COMPARE
192 600 60
$475.60USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, PVDF, Carbon; 0.72 mL/rev EW-07104-59 Mfr # FMI353 / Q2CKY
COMPARE
432 600 60
$468.00USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, Stainless Steel, Carbon; 0.32 mL/rev EW-07104-60 Mfr # FMI359 / Q1CSY
COMPARE
192 600 100
$627.50USD / Each
FMI Series Q Valveless Piston Pump Head, Stainless Steel, Stainless Steel, Carbon; 0.32 mL/rev EW-07104-62 Mfr # FMI363 / Q1SSY
COMPARE
192 600 100
$641.25USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, Stainless Steel, Ceramic; 1.28 mL/rev EW-07104-63 Mfr # FMI462 / Q3CSC
COMPARE
768 600 60
$784.75USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, PVDF, Ceramic; 0.72 mL/rev EW-07104-64 Mfr # FMI355 / Q2CKC
COMPARE
432 600 60
$433.80USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, Stainless Steel, Ceramic; 0.72 mL/rev EW-07104-66 Mfr # FMI212 / Q2CSC
COMPARE
432 600 100
$621.50USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, PVDF, Carbon; 0.72 mL/rev EW-07104-68 Mfr # FMI353 / Q2CKY
COMPARE
432 600 60
$447.90USD / Each
FMI Series Q Valveless Piston Pump Head, Ceramic, Stainless Steel, Carbon; 0.72 mL/rev EW-07104-70 Mfr # FMI360 / Q2CSY
COMPARE
432 600 100
$643.50USD / Each
FMI Series Q Valveless Piston Pump Head, Stainless Steel, Stainless Steel, Carbon; 0.72 mL/rev EW-07104-72 Mfr # FMI364 / Q2SSY
COMPARE
432 600 100
$647.25USD / Each
FMI Series Q Valveless Piston Pump Head, Stainless Steel, PVDF, Carbon; 0.32 mL/rev EW-07117-26 Mfr # FMI361 / Q1SKY
COMPARE
192 600 60
$447.90USD / Each
FMI Series Q Valveless Piston Pump Head, Stainless Steel, PVDF, Carbon; 0.72 mL/rev EW-07117-28 Mfr # FMI362 / Q2SKY
COMPARE
432 600 60
$447.90USD / Each
Showing 1- 17 of 17