Four-Blade Impellers (Lightnin A-200)
Four-blade impellers are turbine impellers that have a 45 degree pitch and produce a semi-axial flow
$425.00 - $544.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Impeller Size (mm) Material Description
Availability Pricing
Lightnin 146835316 316 SS four-blade impeller ( A-200), 3.0" dia x 5/16" bore dia EW-04560-60 Mfr # 146835316
COMPARE
76.2 316 Stainless Steel 316 SS four-blade impeller ( A-200), 3.0\" dia x 5/16\" bore dia
$481.00USD / Each
Lightnin 829448-1 316 SS four-blade impeller ( A-200), 2.0" dia x 3/8" bore dia EW-04560-61 Mfr # 829448-1
COMPARE
50.8 316 Stainless Steel 316 SS four-blade impeller ( A-200), 2.0\" dia x 3/8\" bore dia
$544.00USD / Each
Lightnin 829448 316 SS four-blade impeller ( A-200), 2.0" dia x 5/16" bore dia EW-04561-60 Mfr # 829448
COMPARE
50.8 316 Stainless Steel 316 SS four-blade impeller ( A-200), 2.0\" dia x 5/16\" bore dia
$425.00USD / Each