This product is now discontinued, View Related Products

Gillis Mfg Four-Shelf Units

Unique square post design makes units stronger to hold heavier loads
  • Easily customize your shelving unit—purchase a starter set now and add on to it later.
Units with 36" and 48" shelves can hold up to 1000 lb per shelf; units with 60" shelves hold up to 750 lb per shelf; and units with 72" shelves can hold up to 500 lb per shelf. The open wire shelves give you maximum visibility and let air move around freely, resulting in less accumulation of dust and dirt. Steel shelves feature a double finish for extra protection: They are first coated with electro-zinc ( chromate) plating, then with protoxy—a clear, nontoxic epoxy.
Call For Price

16 variations of this product are available.

See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Capacity (lbs) Number of Shelves Description
Availability Pricing
Gillis Mfg A1836CP64A-4 Add-on shelving unit for four-shelf starter unit 47524-01 EW-47524-11 Mfr # A1836CP64A-4
COMPARE
1000 4 Add-on shelving unit for four-shelf starter unit 47524-01
Call For Price
Gillis Mfg A1848CP64A-4 Add-on shelving unit for four-shelf starter unit 47524-02 EW-47524-12 Mfr # A1848CP64A-4
COMPARE
1000 4 Add-on shelving unit for four-shelf starter unit 47524-02
Call For Price
Gillis Mfg A1860CP64A-4 Add-on shelving unit for four-shelf starter unit 47524-03 EW-47524-13 Mfr # A1860CP64A-4
COMPARE
750 4 Add-on shelving unit for four-shelf starter unit 47524-03
Call For Price
Gillis Mfg A1872CP64A-4 Add-on shelving unit for four-shelf starter unit 47524-04 EW-47524-14 Mfr # A1872CP64A-4
COMPARE
500 4 Add-on shelving unit for four-shelf starter unit 47524-04
Call For Price
Gillis Mfg A2436CP64A-4 Add-on shelving unit for four-shelf starter unit 47524-05 EW-47524-15 Mfr # A2436CP64A-4
COMPARE
1000 4 Add-on shelving unit for four-shelf starter unit 47524-05
Call For Price
Gillis Mfg A2448CP64A-4 Add-on shelving unit for four-shelf starter unit 47524-06 EW-47524-16 Mfr # A2448CP64A-4
COMPARE
1000 4 Add-on shelving unit for four-shelf starter unit 47524-06
Call For Price
Gillis Mfg A2460CP64A-4 Add-on shelving unit for four-shelf starter unit 47524-07 EW-47524-17 Mfr # A2460CP64A-4
COMPARE
750 4 Add-on shelving unit for four-shelf starter unit 47524-07
Call For Price
Gillis Mfg A2472CP64A-4 Add-on shelving unit for four-shelf starter unit 47524-08 EW-47524-18 Mfr # A2472CP64A-4
COMPARE
500 4 Add-on shelving unit for four-shelf starter unit 47524-08
Call For Price
Gillis Mfg A1836CP72A-4 Add-on shelving unit for four-shelf starter unit 47525-01 EW-47525-11 Mfr # A1836CP72A-4
COMPARE
1000 4 Add-on shelving unit for four-shelf starter unit 47525-01
Call For Price
Gillis Mfg A1848CP72A-4 Add-on shelving unit for four-shelf starter unit 47525-02 EW-47525-12 Mfr # A1848CP72A-4
COMPARE
1000 4 Add-on shelving unit for four-shelf starter unit 47525-02
Call For Price
Gillis Mfg A1860CP72A-4 Add-on shelving unit for four-shelf starter unit 47525-03 EW-47525-13 Mfr # A1860CP72A-4
COMPARE
750 4 Add-on shelving unit for four-shelf starter unit 47525-03
Call For Price
Gillis Mfg A1872CP72A-4 Add-on shelving unit for four-shelf starter unit 47525-04 EW-47525-14 Mfr # A1872CP72A-4
COMPARE
500 4 Add-on shelving unit for four-shelf starter unit 47525-04
Call For Price
Gillis Mfg A2436CP72A-4 Add-on shelving unit for four-shelf starter unit 47525-05 EW-47525-15 Mfr # A2436CP72A-4
COMPARE
1000 4 Add-on shelving unit for four-shelf starter unit 47525-05
Call For Price
Gillis Mfg A2448CP72A-4 Add-on shelving unit for four-shelf starter unit 47525-06 EW-47525-16 Mfr # A2448CP72A-4
COMPARE
1000 4 Add-on shelving unit for four-shelf starter unit 47525-06
Call For Price
Gillis Mfg A2460CP72A-4 Add-on shelving unit for four-shelf starter unit 47525-07 EW-47525-17 Mfr # A2460CP72A-4
COMPARE
750 4 Add-on shelving unit for four-shelf starter unit 47525-07
Call For Price
Gillis Mfg A2472CP72A-4 Add-on shelving unit for four-shelf starter unit 47525-08 EW-47525-18 Mfr # A2472CP72A-4
COMPARE
500 4 Add-on shelving unit for four-shelf starter unit 47525-08
Call For Price
Showing 1- 16 of 16

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat