Fowler Dial Indicators

High quality multi-function indicator at a reasonable cost
  • Offers direct inch/metric or inch/metric/fraction conversion
  • Easy-to-read dial face
  • Simple to operate
These economical battery operated indicators offer absolute/incremental readings on a 2 1/4" digital face. Features include tolerance indicators, floating zero, and carbide tipped point.
$120.75 - $203.75USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max range (in) Resolution Description
Availability Pricing
Fowler 54-520-255 Electronic Indicator, 0 to 0.5" (0 to 12.5 mm) EW-10508-51 Mfr # 545202550
COMPARE
0.5 0.0005 in Electronic Indicator; 0 to 0.5"/0 to 12.5mm
$120.75USD / Each
Fowler 54-520-250 Electronic Indicator,  0 to 1" (0 to 25 mm) EW-10508-55 Mfr # 545202500
COMPARE
1 0.0005 in Electronic Indicator; 0 to 1"/0 to 25mm
$179.00USD / Each
Fowler 54-580-250 AGD Indi-X Blue Electronic Indicator/Stand Combo, 0-1" EW-10508-64 Mfr # 545802500 BOM
COMPARE
1 0.0005 in AGD Indi-X Blue Electronic Indicator/Stand Combo; 0-1"
$203.75USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat