Front View Distillation Apparatus, Group 4
  • Observe temperatures of samples when evaporated and condensed under controlled conditions
  • Conforms to ASTM D 86, D 285, and related ASTM international specifications
Front View Distillation Apparatus, Group 4
Designed for testing grade no. 2 fuel oil, grade no. 2-D diesel fuel oil, and other distillates requiring condenser bath temperatures up to 140°F (60°C). Also suitable for gasolines, aviation turbine fuels, napthas, kerosenes and other liquid petroleum products. Features a 300 W copper immersion condenser heater with stepless electronic control.
$5,724.00 - $6,631.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Koehler K45200 Front View Distillation Apparatus, Group 4, right side, 110 VAC EW-59873-20 Mfr # K45200
COMPARE
115
$5,724.00USD / Each
Koehler K45290 Front View Distillation Apparatus, Group 4, right side, 220 VAC EW-59873-25 Mfr # K45290
COMPARE
220
$6,043.00USD / Each
Koehler K45300 Front View Distillation Apparatus, Group 4, left side, 115 VAC EW-59873-30 Mfr # K45300
COMPARE
115
$6,259.00USD / Each
Koehler K45390 Front View Distillation Apparatus, Group 4, left side, 220 VAC EW-59873-35 Mfr # K45390
COMPARE
220
$6,631.00USD / Each