This product is now discontinued, View Related Products
Fuji PXR Process / Temperature Controllers with Fuzzy Logic
The new PXR series controllers are packed with features, to meet a wide variety of needs in the process industries. Low-cost options include RS485 communications, analog retransmission, digital input, timer function and programmable alarms.
One of the most impressive features is the large LED display. The faceplate, designed for NEMA 4X (IP66 equivalent), is watertight and corrosion-resistant. The easy-to-use 3-button keypad allows for easy programming. The screw terminal on the back further reduces the cost by eliminating the need for sockets.
These controllers have all the standard features that you can expect from a standard PID controller and more. In addition to auto-tuning and fuzzy control, they now come with self-tuning – an innovation in the control field. It automatically retunes the controller under certain conditions without the need to revert to auto-tuning. The standard 8-segment ramp/soak feature has been expanded to include two patterns that can be linked to create a 16-step profile. The PXR controllers accept temperature and process inputs, and offer two control outputs and two programmable alarms.
Remote monitoring of up to 31 controllers at a time is possible with the RS485 option that uses the industry-standard Modbus™ protocol; comes with free Windows ® based software. Programs for different applications can be saved to and from the controller.
Call Davis for more details.
Call For Price
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
Compareitem specifications
DIN Size On / Off Function PID Function
Availability Pricing
Fuji PXR3-BEY1-5VRA1 PXR Temp/Process Controller, 1/32DIN; Analog In, 1-Pt High/Low, Retran EW-68457-45 Mfr # PXR3-BEY1-5VRA1
COMPARE
1/32
Call For Price
Fuji PXR3-RAY1-4V0A1 1/32 Dintc Input Relay Outpu EW-68457-46 Mfr # PXR3-RAY2-4V0A1
COMPARE
1/32
Call For Price
Fuji PXR3-RAY1-5V0A1 PXR3RAY15V0A1 1/32 Din Tc Input Relay Outpu 1 Point High/Low Alarm EW-68457-47 Mfr # PXR3-RAY1-5V0A1
COMPARE
1/32
Call For Price
Fuji PXR3-RCY1-4V0A1 1/32 Din Tc Input S Sr/Ssc Output EW-68457-48 Mfr # PXR3-RCY2-4V0A1
COMPARE
1/32
Call For Price
Fuji PXR3-RCY1-5V0A1 PXR3RCY15V0A1 1/32 Dintx Input Ssr/Ssc Outp 1 Point High/Low Alarm EW-68457-49 Mfr # PXR3-RCY1-5V0A1
COMPARE
1/32
Call For Price
Fuji PXR3-REY1-5V0A1 TC input, 4-20 mA DC output, 1-point high/low alarm EW-68457-50 Mfr # PXR3-REY1-5V0A1
COMPARE
1/32
Call For Price
Fuji PXR4-BEY1-GV0A1 1/16DIN Temp/Process Controller; Analog In, 2-Pt High/Low EW-68457-51 Mfr # PXR4-BEY1-GV0A1
COMPARE
1/16
Call For Price
Fuji PXR4-BEY1-GVMA1 PXR 1/16DIN Temp/Process Controller; Analog In, 2-Pt High/Low, RS485 EW-68457-52 Mfr # PXR4-BEY1-GVMA1
COMPARE
1/16
Call For Price
Fuji PXR4-RAY1-4V0A1 Px4RAY14V0A1 1-16 Dintc Input Relay Outpu EW-68457-53 Mfr # PXR4-RAY1-4V0A1
COMPARE
1/16
Call For Price
Fuji PXR4-RAY1-GV0A1 PXR4RAY1GV0A1 1-16 Dintc Input Relay Outpu 2 Point High/Low Alarm EW-68457-54 Mfr # PXR4-RAY1-GV0A1
COMPARE
1/16
Call For Price
Fuji PXR4-RCY1-4V0A1 1-16 Dintc Input Ssr/Ssc Outp 2 Point High/Low Alarm EW-68457-55 Mfr # PXR4-RCY1-4V0A1
COMPARE
1/16
Call For Price
Fuji PXR4-RCY1-GV0A1 TC input, SSR/SSC output, 2-point high/low alarm EW-68457-56 Mfr # PXR4-RCY1-GV0A1
COMPARE
1/16
Call For Price
Fuji PXR5-BEY1-GV0A1 PXR5BEY1GV0A1 1/8 Din 4-20MA/1-5V Dc Inp 2 Point High/Low Alarm EW-68457-57 Mfr # PXR5-BEY1-GV0A1
COMPARE
1/8
Call For Price
Fuji PXR5-RAY1-4V0A1 PXR5RAY14V0A1 1/8 Dintc Input Relay Outpu EW-68457-58 Mfr # PXR5-RAY1-4V0A1
COMPARE
1/8
Call For Price
Fuji PXR5-RAY1-GV0A1 PXR5RAY1GV0A1 1/8 Dintc Input Relay Outpu 2 Point High/Low Alarm EW-68457-59 Mfr # PXR5-RAY1-GV0A1
COMPARE
1/8
Call For Price
Fuji PXR5-RCY1-4V0A1 PXR5RCY14V0A1 1/8 Dintc Input Ssr/Ssc Outp EW-68457-60 Mfr # PXR5-RCY1-4V0A1
COMPARE
1/8
Call For Price
Fuji PXR5-RCY1-GV0A1 PXR5RCY1GV0A1 1/8 Tintc Input Ssr/Ssc Outp 2 Point High/Low Alarm EW-68457-61 Mfr # PXR5-RCY1-GV0A1
COMPARE
1/8
Call For Price
Fuji PXR5-REY1-GV0A1 1/8 Dintc Input 4-20MA Dc Output 2 Point High/Low Alarm EW-68457-62 Mfr # PXR5-REY1-GV0A1
COMPARE
1/8
Call For Price
Fuji PXR9-RAY1-GV0A1 1/4 Dintc Input Relay Outpu 2 Point High/Low Alarm EW-68457-63 Mfr # PXR9-RAY1-GV0A1
COMPARE
1/4
Call For Price
Fuji PXR9-RCY1-4V0A1 PXR9RCY14V0A1 1/4 Dintc Input Ssr/Ssc Outp EW-68457-64 Mfr # PXR9-RCY1-4V0A1
COMPARE
1/4
Call For Price
Showing 1 - 20 of 23