This product is now discontinued, View Related Products
Fuji Strip Chart Recorders - 6 or 12 Channels
Strip Chart Recorders with Brilliant Six-Color Printing
  • Powerful Set of Printing and
  • Recording Features
  • Front Panel Programming
  • Large Fluorescent,
  • Multi-Language Display
  • Non-Contact, Inkjet Printing
  • One Mechanical Assembly for
  • Printing
  • Side-by-Side Mounting
  • Logging Mode
  • Daily Report
  • Zoom, Zone and Auto
  • Range Trace
  • Three-Year Warranty
    Why use dot-matrix or fiber-tip pen printing when this cutting-edge recording technology offers an affordable inkjet recorder? In addition, this Series has an excellent rating for measuring range accuracy and analog trace accuracy, and carries a three-year warranty.
    The 100mm and the 180mm recorders offer brilliant six color recording. Tiny piezo elements electronically
    dispense small dots of colored ink, 0.3mm in diameter, onto the chart paper. This non-contact printing advancement traces vibrant trend lines and prints sharp alphanumeric characters. This technology, previously available on expensive printers only, now comes in a strip chart
    recorder at an affordable price. If you note the difference in print quality between the dot-matrix and the inkjet typefaces, you’ll conclude there’s simply no reason to use a dot-matrix type recorder anymore.
    The inkjet cartridges have a long life of up to 6 months of continuous operation. Each channel is assigned its own color, and reports generated in reference to that channel are printed in the corresponding color. As
    this method of printing requires only one mechanism, these recorders require fewer moving parts than conventional recorders - that means years of extremely reliable, trouble-free operation for you.
    As each channel has its own 16-bit A/D converter, these instruments offer high speed sampling of the input signal. Furthermore, these recorders are fully programmable from their easy-to-read, fluorescent front panels for a variety of parameters such as:
    • Pass code
    • Main and Sub chart speeds
    • Record mode - trend/logging
    • Recording range
    • Input signal
    • Tag number
    • Communication parameters
    • Ink monitor status
    • Language (English, French, or German)
    • List print request
    • Daily report, date and time, and alarm settings
    • Illumination status
    • And more…
  • Call For Price
    2 variations of this product are available.
    See All Product Options Below
    2 PRODUCT OPTIONS
    Compareitem specifications
    Chart Diameter (in) Chart Speed Max Input (VDC)
    Availability Pricing
    Fuji PHA98004-EA0YV Strip Chart Recorder, 180 mm wide, 12-channel EW-49002-25 Mfr # PHA98004-EA0YV
    COMPARE
    3 to 1500 mm/hr (user selectable)
    Call For Price
    Fuji PHC66003-AA0YV Strip Chart Recorder, 100 mm wide, 6-channel EW-49002-26 Mfr # PHC66003-AA0YV
    COMPARE
    5 to 1500 mm/hr (user selectable)