Gas Monitor Calibration
Protect personnel from gas hazards due to inaccurate readings – calibrate your gas detector today!
  • Factory authorized service center for MSA and Sperian gas detectors
InnoCal's specially trained technicians test your gas detector with a known concentration of gas. Calibration drift can occur due to many factors: normal degradation of sensors over time, exposure to extreme temperature and humidity conditions, exposure to high levels of the target gases, and harsh storage and handling conditions such as being dropped or submerged in liquid. Ensure worker safety by having your gas detector calibrated by our factory-trained technicians to verify proper operation.
$220.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Calibration Type Number Of Calibration Test Points Description
Availability Pricing
COMPARE
Air and Gas Measurement 1 NIST-Traceable calibration for carbon dioxide meters
$220.00USD / Each
COMPARE
Air and Gas Measurement 1 NIST-Traceable calibration for oxygen meters
$220.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat