GE Druck High-Accuracy Industrial Pressure Transmitters
All-stainless-steel construction for heavy industrial use
  • ±0.1% full-scale accuracy
These high-accuracy transmitters combine proven silicon technology and analog circuitry with a compact design. The Druck silicon technology is at the heart of the transmitter, resulting in a range of high quality piezo-resistive pressure sensors which are both robust and stable. High levels of performance have been achieved without the use of digital electronics. These transmitters are particularly well suited to sensitive applications, such as those requiring low power, low noise, pulse power, and fast response.
High-grade stainless steel body and analog electronics provide fast turn on, pulse power, 5 kHz frequency response, and low noise. Additionally, transmitters feature high over-pressure capability, stability typically at ±0.05%, and FM (US and Canada) intrinsically safe certifications.
Higher pressure range transmitters come standard with sealed diaphragm to provide extra pressure equipment safety. Other process and electrical connections are available; contact an Application Specialist for details.
$324.00 - $465.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (PSI)
Availability Pricing
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 1/4" NPT(F), 15 to 30 psig EW-98071-00 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 30
$324.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 1/4" NPT(F), 11.5 to 17 psi barometric EW-98071-01 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability)
$324.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 1/4" NPT(F), 0 to 5 psig EW-98071-02 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 5
$324.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 1/4" NPT(F), 0 to 50 psig EW-98071-03 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 50
$324.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 1/4" NPT(F), 0 to 100 psig EW-98071-04 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 100
$324.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 1/4" NPT(F), 0 to 200 psig EW-98071-05 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 200
$324.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 1/4" NPT(F), 0 to 300 psig EW-98071-06 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 300
$324.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 1/4" NPT(F), 0 to 500 psig EW-98071-07 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 500
$324.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 1500 psi, Sealed EW-98071-08 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 1500
$385.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 3000 psi, Sealed EW-98071-09 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 3000
$465.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 5000 psi, Sealed EW-98071-10 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 5000
$385.00USD / Each
GE Druck PTX5072-TC-A2-CA-H6-PE High-Accuracy Pressure Transmitter, 10000 psi, Sealed EW-98071-11 Mfr # PTX5072-TC-A2-CA-H6-PE
COMPARE
±0.1% full-scale (combined effects of nonlinearity, hysteresis, and repeatability) 10000
$385.00USD / Each
Showing 1- 12 of 12