This product is now discontinued, View Related Products
GE Druck PTX600 Precision Pressure Transmitters
Designed for critical applications in industrial and research environments, the Druck PTX 600 series of two-wire, 4-20mA output pressure transmitters offer a new level of stability, thermal performance, versatility and measurement accuracy.
A high level of protection against electrical interference has been combined with surface mount components and total potting of the electronics to ensure integrity under high levels of shock and vibration.
The PTX 600 Series features high accuracy, 0.08% F.S. Best Straight Line for the combined effects of non-linearity, hysteresis and repeatability, and to fully represent operating conditions, the thermal effects are expressed as a maximum change in output from the calibration at room temperature.
The transmitter provides an integral 1/4” NPT female pressure port and a screw-in stainless steel male to male adapter available as an option.
Comes with one screw-in 1/4 NPT male/male adapter FEATURES
Call For Price
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
GE Druck PTX 610 Pressure Transmitter, 5PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-86 Mfr # PTX 610 5PSIG
COMPARE
±0.08% full-scale 5
Call For Price
GE Druck PTX 610 Pressure Transmitter, 100PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-87 Mfr # PTX 610 100PSIG
COMPARE
±0.08% full-scale 100
Call For Price
GE Druck PTX 610 Pressure Transmitter 10000 PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-88 Mfr # PTX 610 10000PSIG
COMPARE
±0.08% full-scale 10000
Call For Price
GE Druck PTX 610 Pressure Transmitter 11.5/17PSI Intrinsically Safe, 0.08%FS Accuracy EW-68441-89 Mfr # PTX610-1176-(11.5/17PSIA)
COMPARE
±0.08% full-scale
Call For Price
GE Druck PTX 610 Pressure Transmitter, 15PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-90 Mfr # PTX 610 15PSIG
COMPARE
±0.08% full-scale 15
Call For Price
GE Druck PTX 610 Pressure Transmitter 1500PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-91 Mfr # PTX 610 1500PSIG
COMPARE
±0.08% full-scale 1500
Call For Price
GE Druck PTX 610 Pressure Transmitter -15/30PSI Intrinsically Safe, 0.08%FS Accuracy EW-68441-92 Mfr # PTX610-1176-(-15/30PSIG)
COMPARE
±0.08% full-scale 30
Call For Price
GE Druck PTX 610 Pressure Transmitter 200PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-93 Mfr # PTX 610 200PSIG
COMPARE
±0.08% full-scale 200
Call For Price
GE Druck PTX 610 Pressure Transmitter 300PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-94 Mfr # PTX 610 300PSIG
COMPARE
±0.08% full-scale 300
Call For Price
GE Druck PTX 610 Pressure Transmitter 3000PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-95 Mfr # PTX 610 3000PSIG
COMPARE
±0.08% full-scale 3000
Call For Price
GE Druck PTX 610 Pressure Transmitter, 50PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-96 Mfr # PTX 610 50PSIG
COMPARE
±0.08% full-scale 50
Call For Price
GE Druck PTX 610 Pressure Transmitter 500PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-97 Mfr # PTX 610 500PSIG
COMPARE
±0.08% full-scale 500
Call For Price
GE Druck PTX 610 Pressure Transmitter 5000PSIG Intrinsically Safe, 0.08%FS Accuracy EW-68441-98 Mfr # PTX 610 5000PSIG
COMPARE
±0.08% full-scale 5000
Call For Price
Showing 1- 13 of 13