This product is now discontinued, View Related Products
GE (General Eastern) Industrial Relative Humidity/Dew Point Transmitter
Ideal for low or no-flow applications
  • Versatile analyzers measure RH, dew point, absolute humidity, and mixing ratio
Analyzers are designed to be inserted into pipes or vessels and secured with the 1/2" NPT(M) fitting. They eliminate the need for a separate sampling pump.
Analyzer head features epoxy-coated cast aluminum construction, meeting NEMA 4X, IP66 requirements. Models with an integral display provide local indication of process variables and on-site setup via the touch pad. All models have two-wire connections to connect with 4 to 20 mA output devices for data display, storage, and/or analysis. Single- and dual-output models are available.
Probe features 316 stainless steel construction with a fast-response, silicon-based polymer sensing element. To help prevent contamination and premature clogging, the sensing element is fitted with a 100-micron sintered filter protective cap.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Display Type Max RH Range (%) Mounting Type
Availability Pricing
General Eastern MR2350 Industrial Relative Humidity/Dew Point Analyzer, One Output EW-37405-00 Mfr # MMR31-R3A1A
COMPARE
100
Call For Price
General Eastern MR2350 Industrial RH//Dew Point Analyzer; Display/1-Output EW-37405-04 Mfr # MMR31"A"-R-3-A-1-G
COMPARE
LCD 100
Call For Price
General Eastern MR2350 Industrial RH//Dew Point Analyzer; Display/2-Output EW-37405-06 Mfr # MMR31"A"-R-3-A-1-H
COMPARE
LCD 100