This product is now discontinued, View Related Products
GE Kaye Ice Point and Uniform Temperature Reference
Obtaining high-accuracy measurements from thermocouples requires instrumentation designed to minimize errors. Thermocouple conformity, the cold junction and its reference, the A/D converter and other analog system electronics all contribute to the overall system error. While the cold junction (termination of thermocouple to copper) can contribute significantly to system error, this error can be minimized with a high accuracy temperature reference.
Uniform Temperature References
The GE Kaye Uniform Temperature References (UTRs) provide cold junction temperature measurement for up to 64 thermocouples with twice the accuracy of conventional models. This high level of accuracy is achieved through remove monitoring via a NIST-traceable RTD, which automatically performs compensation calculations. The UTRs are passive mechanical devices requiring no external power.
Operates in temperature extremes
Easily reconfigured for different thermocouple types
Can be installed near test article in the test environment
Ice Point References
GE Kaye's Ice Point References provide the highest level of accuracy in automatic referencing - zero long-term drift maintaining reference temperature at 0 °C. Ice Point References are available in rack-mount or portable configurations with capacity for up to 75 thermocouples.
  • Operates from 0 to 40 °C
  • Negligible effect from rapid changes in ambient
  • Convenient for more permanent installations
  • Limited to stable areas of vibration and ambient, e.g., control room
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy Temperature Resolution (° F)
Availability Pricing
Amphenol X0240 GE Advanced Sensors Auto Ice Point Bath EW-16101-29 Mfr # X0240
COMPARE
Call For Price
Amphenol X0262 Ice Point Reference, 24 Channel SS880A-0-0-0-0 EW-16101-30 Mfr # 2083
COMPARE
Call For Price
Amphenol X0264 GE Advanced Sensors Ice Point Reference, 50 Channel EW-16101-31 Mfr # X0264
COMPARE
Call For Price
Amphenol X0265 GE Advanced Sensors Ice Point Reference, 75 Channel EW-16101-32 Mfr # X0265
COMPARE
Call For Price
Kaye X0280 Temperature Reference, UTR-AS Shielded Plate, 32 Channel with Enclosure EW-16101-33 Mfr # X0280
COMPARE
Call For Price
Kaye X0290 Temperature Reference; UTR-AS Non-Shield/48 Channel/Enclosure EW-16101-34 Mfr # X0290
COMPARE
Call For Price
Kaye V0360 Calibrated Resistance Temperature Detector EW-16101-35 Mfr # V0360
COMPARE
Call For Price
Kaye V0370 4-Wre Platinum Element EW-16101-36 Mfr # V0370
COMPARE
Call For Price
Showing 1- 8 of 8