GE Panametrics Accessories for Flowmeters

Keep your GE Panametrics flowmeter running accurately with these parts and accessories.
$225.00 - $1,710.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GE Panametrics Transducer 2MHZ 2-8 inch pipe EW-32486-76 Mfr # UTXDR-407-3C-25-00-IN-0
COMPARE
Transducer 2MHZ 2-8 inch pipe
$1,362.00USD / Each
GE Panametrics UTXDR Small Pipe Transducer, 0.5 to 2", 4 MHz EW-32487-06 Mfr # UTXDR-408-LM-25-00 IN-0
COMPARE
Small Pipe Transducer, 0.5 to 2", 4 MHz
$1,480.00USD / Each
GE Panametrics 234-024 IRDA Communication Cable EW-68453-01 Mfr # 234-050
COMPARE
IRDA Communication Cable
$225.00USD / Each
GE Panametrics C-RS-402-00-0 Transducer Pair, for 2 to 24" Pipe, 1.0MHz EW-68453-02 Mfr # C-RS-402-00-0
COMPARE
Transducer Pair, for 2 to 24" Pipe, 1.0MHz
$1,710.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat