GE Sensing Duct and Wall Mount CO2 Sensors
FEATURES
  • Gas sensing engine provides high accuracy in a compact low cost package
  • ABC Logic™ self-calibration system eliminates the need for manual calibration in most applications
  • Gold plated optical sensor increases sensor life and durability
  • Mounting bracket with terminal block provides quick, easy wiring
  • Dual simultaneous analog output (V & mA)
  • On board relay with adjustable set point and dead band
  • Choice of 9 preprogrammed ''standard settings'' on easy to use interface
  • PC interface and adjustable settings allows for simple configuraiton
  • Optional enclosures available
  • Lifetime calibration guarantee
$219.00 - $329.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max CO2 (ppm) Max CO2 (%)
Availability Pricing
GE 8001B CO2 Sensor No Display Black Ventostat Wall Mount EW-10101-42 Mfr # T8100B
COMPARE
$219.00USD / Each
Amphenol 8007 CO2 Sensor with Display, Duct Mount EW-10101-47 Mfr # T8300-DB
COMPARE
$329.00USD / Each