GE (Telaire) Dual-Position CO2/Temperature Meter
  • Identify areas with low or substandard ventilation—take readings in less than 60 seconds! Measure CO2 levels up to 10,000 ppm Dual-beam infrared absorption gas sensor provides long term stability and durability Use with batteries for mobility or use the included AC/DC adapter for local monitoring Maintain CO2 accuracy by recalibrating on site—use the optional calibration kit sold separately Easily adjusted for changes in altitude
Dual-beam infrared absorption gas sensor provides long term stability and durability. Also features a dual display, analog output, and a flip-out stand for desktop monitoring. Maintain CO2 accuracy by recalibrating on site—use the optional calibration kit sold separately.
$606.25 - $661.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Parameters Measured
Availability Pricing
Modus 7001 Dual-Position CO2 and Temperature Meter EW-86290-00 Mfr # T7001
COMPARE
CO2, Temperature
$606.25USD / Each
Modus 7001D Dual Position CO2, Temperature, and Humidity Meter EW-86290-10 Mfr # T7001D
COMPARE
CO2, Temperature, Humidity
$661.00USD / Each