Gems LS-3 Series Level Switches
Install from the top of your tank—attach your own conduit to protect wiring.
  • Get longer life and more durability with hermetically sealed switches
All models accept 120/240 VAC and operate as NO or NC switches except where noted. Switches include 24" leads (except 07187-12 which has 72" leads). If your level control system exceeds the current rating in the table, use a controller with your switch.
$46.35 - $201.20USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Min Secific Gravity Description
Availability Pricing
Gems LS-3 Level Switch, Polypropylene; 50 psig, 3/8" Straight Thread EW-07187-02 Mfr # 171517
COMPARE
50 0.6 LS-3 Level Switch, Polypropylene; 50 psig Max Pressure, 3/8" Straight Thread
$69.55USD / Each
Gems LS-3 Level Switch, Polypropylene, 50 psig Max Pressure, 1/8" NPT EW-07187-04 Mfr # 142505
COMPARE
50 0.6 LS-3 Level Switch, Polypropylene, 50 psig Max Pressure, 1/8" NPT
$46.35USD / Each
Gems Standard float switch; polypropylene (solid), 3/8"-16 thread mounting EW-07187-06 Mfr # 171514
COMPARE
150 0.9 Standard float switch; polypropylene (solid), 3/8"-16 thread mounting
$49.25USD / Each
Gems LS-3 Level Switch, Polypropylene; 150 psig Max Pressure, 1/8 NPT EW-07187-08 Mfr # 142545
COMPARE
150 0.9 LS-3 Level Switch, Polypropylene; 150 psig Max Pressure, 1/8 NPT
$52.30USD / Each
Gems Standard Level Switch, Polypropylene, 0.90 Minimum Specific Gravity, 1/8" NPT EW-07187-10 Mfr # 116826
COMPARE
150 0.9 LS-3 Level Switch, Polypropylene Float 0.90 Minimum Liquid Specific Gravity, 1/8" NPT
$46.35USD / Each
Gems Standard float switch; SS with slosh shield, (2) 9/32" mounting holes EW-07187-12 Mfr # 43765
COMPARE
150 0.55 Standard float switch; SS with slosh shield, (2) 9/32" mounting holes
$201.20USD / Each
Gems LS-3  Standard Level Switch, PVDF, 0.86 Minimum Specific Gravity, 1/8" NPT EW-07187-14 Mfr # 173250
COMPARE
50 0.86 LS-3 Level Switch, PVDF Float 0.86 Min Liquid Specific Gravity, 50 psi Max Pressure
$75.00USD / Each
Gems Standard float switch; brass, 1/8" NPT mounting EW-07187-16 Mfr # 01701
COMPARE
150 0.45 Standard float switch; brass, 1/8" NPT mounting
$76.50USD / Each
Gems LS-1700 Series Level Switch, 316ss Stem Buna-N Float 20VA EW-07187-18 Mfr # 01702
COMPARE
150 0.45 LS-1700 Series Level Switch, 316ss Stem Buna-N Float 20VA
$135.00USD / Each
Gems Standard float switch; stainless steel, 1/8" NPT mounting EW-07187-20 Mfr # 79990
COMPARE
100 0.7 Standard float switch; stainless steel, 1/8" NPT mounting
$148.90USD / Each
Gems Standard float switch; CPVC, 1/4" NPT mounting EW-07188-40 Mfr # 74780
COMPARE
15 0.85 Standard float switch; CPVC, 1/4" NPT mounting
$101.70USD / Each
Showing 1- 11 of 11