General Tools Digital Sound Level Meter with Data Logging SD Card
Store data in Excel® format on the SD card
  • Performs automatic data logging at sampling time settable from 1 second to 1 hour Tripod mountable—ideal for field use Easy-to-read, large 2-1/2” diagonal backlit LCD
Use these sound meters to accurately measure the noise level of an environment or a piece of machinery to help comply with OSHA guidelines. Meters allow you to store sampled data in Excel format on SD cards so you can easily view and graph the collected data. Measure from 30 to 130 dB in auto-ranging mode with A or C weighting mode in fast/slow response time. Additional features include Min/Max/peak and data Hold functions, an RS-232 port, and a flip-up stand.
$469.00 - $573.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Meter Class
Availability Pricing
General Tools & Instruments DSM402SD Digital Class 2 Sound Level Meter with Data Logging SD Card EW-37800-02 Mfr # DSM402SD
COMPARE
Class 2
$469.00USD / Each
General Tools & Instruments DSM403SD Digital Class 1 Sound Level Meter with Data Logging SD Card EW-37800-03 Mfr # DSM403SD
COMPARE
Class 1
$573.00USD / Each
Showing 1- 2 of 2
Need Help with this product?
Call Us At1-847-549-7600
Live chat