General Tools Infrared Thermometer with Star Burst Laser
Tri-color light bar and three-speed buzzer indicate if temperature is within or outside set range
  • Unique STAR BURST laser targeting system
  • Fast response time of 0.5 seconds
Ideal for conducting temperature audits. Infrared thermometer progressively scans an area and checks for temperature differentials which indicate changes in temperature. The “star burst” laser targeting system helps you determine the approximate target size and location, reducing possible false alarms. Features include a large backlit LCD, Min/Max, data hold, and autopower off.
ITEM(S) ON SALE
$67.15 - $91.80USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Max Temperature (° F)
Availability Pricing
General Tools IRTC50 Seeker Infrared Thermometer and Scanner with Star Burst Laser Target (8: 1) EW-37803-69 Mfr # IRTC50
COMPARE
8:1 Fixed 428
$67.23USD / Each
$79.10USD / Each
General Tools IRT657 Seeker Infrared Thermometer with Star Burst Laser Target (12: 1) EW-37803-74 Mfr # IRT657
COMPARE
12:1 Fixed 1076
$67.15USD / Each
$79.00USD / Each
General Tools IRT659K Seeker Infrared Thermometer and Scanner with Star Burst Laser Target (12: 1) EW-37803-75 Mfr # IRT659K
COMPARE
12:1 Adjustable 1202
$91.80USD / Each
$108.00USD / Each