General Tools Pin/Pinless Digital Moisture Meters with Thermohygrometer
Stores up to 20 pairs of temperature and RH readings
  • Settable high and low alarms for RH and moisture level Easy to read backlit LCD with 3-digit primary readout Auto power off function saves battery life
Combination pin/pinless moisture meters offer temperature and humidity readings and additional calculations, making them ideal for water damage remediators and facility maintenance professionals. Each unit features a data Hold function, Min/Max values, and temperature units in °F or °C.
ITEM(S) ON SALE
$258.82USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
General Tools RHMG650 9-in-1 Pin/Pinless Digital Moisture Meter with Thermohygrometer EW-37800-16 Mfr # RHMG650
COMPARE
9-in-1 Pin/Pinless Digital Moisture Meter with Thermohygrometer
$258.82USD / Each
$304.50USD / Each