Jenway® Genova Nano Micro-Volume UV/Visible Spectrophotometer
Measure sample volumes as low as 0.5 μL with a high degree of accuracy and reproducibility in less than 6.5 seconds!
  • Preprogrammed methods for DNA, RNA and Protein measurements
  • Detects DNA concentrations as low as 2 ng/μL
  • Icon-driven software and easy-to-use navigation system provide intuitive usability
  • Method and result saving to USB memory stick
  • Easy and quick to clean
Measure sample volumes as low as 0.5 µL with the Genova Nano spectrophotometer. Low-volume sample helps conserve precious samples, reduces the need for dilutions, and eliminates the requirement for cuvettes.
The versatile Genova Nano spectrophotometer has all the features of a standard spectrophotometer plus features that are dedicated to life science analysis. Preprogrammed methods measure ssDNA, dsDNA, RNA, and oligonucleotide concentrations as well as Pierce 660, Bradford, Lowry, Biuret, and Bicinchoninic Acid (BCA) protein analysis. Measure DNA concentrations and purity ratios using wavelengths recorded at 260, 280 and 230 nm, with an optional correction at 320 nm. As a standard spectrophotometer, measurement modes include photometrics, concentration, multi-wavelength, spectrum scanning, quantitation, and kinetics.
The stainless steel read head with an embedded quartz lens uses the natural surface tension of the droplet to form the bond between the read head surfaces. To clean, simply wipe the read heads with a microfiber cloth to remove all trace of the sample, allowing faster change over between samples and therefore increasing sample throughput.
$8,404.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bandwidth Max Wavelength (nm) Min Wavelength (nm)
Availability Pricing
COMPARE
5.0 nm 1000 198
$8,404.00USD / Each