+GF+ Piping Systems Metering Ball Valves
Precise control over 180° range
  • Choose between PVC or polypropylene (PP) construction
Ball valves give you precise control over the entire 180°> range. True union design lets you remove wetted parts. Valves have Viton® or EPDM body seals and seats. Less than 5" overall length. Calibrated in 5°> increments.
$126.70 - $223.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Port Size (NPT(F)) Wetted Materials
Availability Pricing
GF Signet 161-323-551 Piping System Metering Ball Valve, PVC, 3/8" NPT(F)
Always in Stock
EW-01360-40 Mfr # 161523521
COMPARE
0.375 PVC
$126.70USD / Each
GF Signet 161-523-522 Piping System Metering Ball Valve, PVC, 1/2" NPT(F)
Best Seller Products
EW-01360-41 Mfr # 161523522
COMPARE
0.5 PVC
$130.30USD / Each
GF Signet 167-323-412 Piping System Metering Ball Valve, Polypropylene, 1/2" NPT(F) EW-01360-45 Mfr # 167523412
COMPARE
0.5 Polypropylene
$223.50USD / Each