GF® Signet 316 SS Socket-Weld Mini-Tap Installation Fittings
Installation fittings for your GF® Signet Paddle Wheel Flowmeters
  • 316 SS Socket-Weld Mini-Tap Fittings for Metalex™ sensors only
  • Includes cap kit
Fittings are precision crafted to ensure proper sensor insertion depth and accurate flow measurement. Pipe fitting sizes range from 1/2" to 1" in diameter.
Required system components are the flow sensor, installation fitting, and monitor or controller, all sold separately.
$635.00 - $778.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Pipe Size Wetted Materials
Availability Pricing
GF Signet P526-2005 316 SS Socket-Weld Mini-Tap Fitting; 1/2" Pipe ID EW-05618-61 Mfr # P526-2005
COMPARE
0.5" 316 Stainless Steel
$635.00USD / Each
GF Signet P526-2007 316 SS Socket-Weld Mini-Tap Fittings; 3/4" Pipe ID EW-05618-62 Mfr # P526-2007
COMPARE
0.75" 316 Stainless Steel
$635.00USD / Each
GF Signet P526-2010 316 SS Socket-Weld Mini-Tap Fittings; 1" Pipe ID EW-05618-63 Mfr # P526-2010
COMPARE
1" 316 Stainless Steel
$778.00USD / Each
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat