GF Signet® Manual Diaphragm Valves, PVC

Optimized valve body provides double the CV values
  • Two-color positition indicator on handwheel
  • Elimination of bolts removes corrosive weak points
  • Mount vertically or horizontally
  • PVC body and EPDM diaphragm
These GF Signet® manual diaphragm valves are constructed with a PVC body and EPDM diaphragm. Valves offer a maximum temperature of 150°F (65°C) and feature true union socket connections.
$224.00 - $367.50USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Cv Inlet Port Outlet Port
Availability Pricing
GF Signet 161.514.614 Type 514 PVC Manual Diaphragm Valve, 1", 32.5 Cv EW-07390-50 Mfr # 161.514.614
COMPARE
32.5 1" true union socket 1" true union socket
$224.00USD / Each
GF Signet 161.514.615 Type 514 PVC Manual Diaphragm Valve, 1-1/4" EW-07390-52 Mfr # 161.514.615
COMPARE
51.3 1-1/4" true union socket 1-1/4" true union socket
$316.10USD / Each
GF Signet 161.514.616 Type 514 PVC Manual Diaphragm Valve, 1-1/2" EW-07390-54 Mfr # 161.514.616
COMPARE
85.3 1-1/2" true union socket 1-1/2" true union socket
$342.90USD / Each
GF Signet 161.514.617 Type 514 Manual Diaphragm Valve, PVC, 2" True Union Socket EW-07390-56 Mfr # 161.514.617
COMPARE
116.8 2" true union socket 2" true union socket
$367.50USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat