+GF+ Signet Rotor-X Paddle-Wheel Flow Sensors
Open-core design ensures accurate, linear output to ±1%
Sensors contain an open-core rotor with a small magnet in each of the four blades. As they rotate, the magnets pass a coil in the sensor. The transducer generates a linear frequency output that is proportional to the flow velocity. This signal travels up to 200 feet without amplification.
Open-core design eliminates cavitation at flow velocities from 1 to 20 ft/second. Closed or solid paddle-wheel designs create air bubbles at these velocities, causing a nonlinear, nonrepeatable signal.
Sensor materials resist most chemicals. All models have PVDF rotor and VITON® O-ring; and include a 25-ft connecting cable.
ROTOR-X flow sensors are compatible with all of our line-, battery-, and sensor-powered flowmeters, accumulators, and controllers. Mount in any type of pipe using an installation fitting—order separately.
$572.00 - $993.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Pipe Size Wetted Materials
Availability Pricing
GF Signet 5-51530-PO Rotor-X High Flow Sensor for 1/2 to 4" Pipe, PP Body, Titanium Shaft EW-05618-10 Mfr # P51530-P0
COMPARE
1/2 to 4" Polypropylene, Titanium
$572.00USD / Each
GF Signet P51530-P1 Rotor-X High Flow Sensor for 5" to 8" Pipe, PP Body, Titanium Shaft EW-05618-11 Mfr # P51530-P1
COMPARE
5 to 8" Polypropylene, Titanium
$615.00USD / Each
GF Signet P51530-VO Rotor-X High Flow Sensor for 1/2 to 4" Pipe, PVDF Body, Hastelly C Shaft EW-05618-13 Mfr # P51530-VO
COMPARE
1/2 to 4" PVDF, Hastelloy C
$993.00USD / Each
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat