GFG Fixed Single-Point Gas Monitors/Transmitters
Simple and effective
  • AutoCal® automatic calibration Smart Sensor technology Digital display/control connection Internal push button alarm settings
The Dynagard accurately displays values on a large backlit LCD, and can transmit a linear 4 to 20 mA output. AutoCal® calibration and maintenance is done effortlessly through front panel push-buttons. Smart sensors enable the user to change measuring ranges or gas types in the field without the need for additional programming. The Dynagard is equipped with onboard relays for two adjustable alarm points, and one sensor fault for local connection and activation or deactivation of external devices like strobes, fans, auto dialers, etc.
$550.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Gases Measured
Availability Pricing
GFG Instrumentation 7771-038 Gas Monitor/Transmitter Calibration Kit; Without Gas EW-10109-51 Mfr # 7771-038
COMPARE
O2/Toxic Gasses
$550.00USD / Each