Gilmont Accucal Flowmeters 1733
Use correlated or direct-reading scales—one meter for many applications
  • Direct reading scales in metric or English units—customize the meter
  • for each application
  • Turn-down ratio of greater than 25:1 for outstanding monitoring and control
  • Rugged body with protective shield—ideal for industrial and laboratory use
Each flowmeter package includes correlation charts plus two direct-reading scales. Attach either of the interchangeable direct reading scales for water or air; you may also use the air and water correlation charts along with readings off the flow tube to determine flow rates.
Excellent accuracy is achieved using second-generation correlation techniques along with factory calibration of the tube. This allows for accuracy to be expressed as a percent of reading--not of full-scale! See details below. In addition, all flowtubes are serial numbered for traceability calibration, accuracy, and manufacturing data.
Meters are available in 65-mm and 150-mm scale lengths so users have a choice of either a compact installation or a larger, more precise and readable scale. The polycarbonate shield wraps around 150° of the flowmeter for better viewing. Models with valves feature a 15-turn shallow taper plug design for precise control across 80-plus % of the scaled range.
Meters are designed for easy flowtube exchange or replacement. Along with a snap-off shield, the end stop has a large diameter bore for centering the flowtube more quickly and accurately than other flowmeter designs. All flowtubes are interchangeable within the same frame size.
These meters are suitable for panel or bench mounting on a stand.
ITEM(S) ON SALE
$479.00 - $911.00USD / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Fluid Type Max Flow Rate (mL/min)
Availability Pricing
COMPARE
Water 4.9
$796.00USD / Each
COMPARE
Water 15
$632.00USD / Each
COMPARE
Water 360
$695.00USD / Each
COMPARE
Water 1400
$694.00USD / Each
COMPARE
Water 4.9
$694.00USD / Each
COMPARE
Water 120
$532.00USD / Each
COMPARE
Water 360
$707.00USD / Each
COMPARE
Water 2200
$796.00USD / Each
COMPARE
Water 4700
$752.00USD / Each
COMPARE
Water 4.9
$674.00USD / Each
COMPARE
Water 15
$546.00USD / Each
COMPARE
Water 120
$621.00USD / Each
COMPARE
Water 360
$595.00USD / Each
COMPARE
Water 1400
$569.00USD / Each
COMPARE
Water 4.9
$498.00USD / Each
COMPARE
Water 15
$480.00USD / Each
COMPARE
Water 55
$494.00USD / Each
COMPARE
Water 360
$479.00USD / Each
COMPARE
Water 2200
$911.00USD / Each
COMPARE
Water 4700
$880.00USD / Each
Showing 1 - 20 of 24