Gilmont Micrometer Dispensers
Dispense small volumes at constant velocity—ideal for titrations and fluid injections
Turn the handle of this dispenser for precise delivery of small volumes. Dispense fluids with an accuracy of ±0.5% of reading or ±1 scale division, whichever is greater. Fluids only come in contact with the borosilicate glass tip, PTFE syringe barrel, and Viton® O-rings.
Autoclavable
Syringe-style pipettes terminate in a standard luer-lock fitting for easy replacement and interchangeability. Dispensers include one tip.
$288.20 - $295.90USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
COMPARE
0.2
$295.90USD / Each
COMPARE
2
$288.20USD / Each