This product is now discontinued, View Related Products
GK Heller 10:1 Gear-Reduction Mixers; 2 to 250 rpm (115 VAC, 50/60 Hz)
Maintain constant speed and torque throughout entire speed range
  • X-Y recorder output for charting and calibrating speed and torque
Electronically monitor and record speed and torque. These mixer systems are ideal for polymer chemistry—gather data on viscosity profiles, shear rate and shear stress, starting torque for plastic materials, flocculation points, and more.
All mixers come with a power supply/controller that provides filtered DC power for the motor, changing the voltage as necessary. Maintain constant shaft speed and torque within ±1% of preset values under varying line voltage conditions. Obtain torque readings with a simple flip of a switch.
Permanent magnet, ball-bearing, totally enclosed DC motor has oversized brushes and skewed armature to avoid cogging at low speeds. A fast-acting circuit breaker protects components even if the motor is stalled.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Motor hp
Availability Pricing
GK Heller HST20 10:1 gear-reduction mixer with speed/torque readout; 336 in-oz (max), 115 VAC EW-04407-00 Mfr # 099D HST20
COMPARE
250 336 1/8
Call For Price
GK Heller HST3010 10:1 gear-reduction mixer with speed/torque readout; 720 in-oz (max), 115 VAC EW-04407-20 Mfr # 099D HST3010
COMPARE
250 720 1/4