Glas-Col Flexible Fabric Sixty-Degree Funnel Heating Mantles
Get better heating with conforming flexible fabric
Heating Mantles feature insulated glass fabric that conforms to unusual vessel shapes. Sixty-Degree Funnel Mantles prevent precipitation and reduces viscosity of solutions during transfer. Mantles require a controlled electrical power source to prevent overheating
$496.00 - $555.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
Glas-Col 100AO719 60° Heating Mantle with 6" Top Diameter, 120 Watts, 230 V EW-36227-47 Mfr # 100AO719
COMPARE
230 60° Heating Mantle with 6" Top Diameter, 120 Watts, 230 V
$555.00USD / Each
Glas-Col 100AO718 60° Heating Mantle with 6" Top Diameter, 120 Watts, 115 V EW-36227-56 Mfr # 100AO718
COMPARE
115 60° Heating Mantle with 6" Top Diameter, 120 Watts, 115 V
$496.00USD / Each
Glas-Col 100AO722 60° Heating Mantle with 8" Top Diameter, 210 Watts, 115 V EW-36227-58 Mfr # 100AO722
COMPARE
115 60° Heating Mantle with 8" Top Diameter, 210 Watts, 115 V
$519.00USD / Each