GoatThroat Pump Accessories
Enhance versitility of your GoatThroat™ Spillproof Barrel Pump
Accessories for GoatThroat Spillproof Barrel Pump optimize performance or suitable for maintenance.
The pneumatic air adapter, model 75650-40, allows GoatThroat™ pumps to be used with shop air, air compressors, or bottle compressed air. Uses up to 4 psi to deliver fluids up to 4.5 GPM. External pressure valve keeps containers from being over pressurized.
$43.00 - $208.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Lab/shop air adapter for pumps
$208.00USD / Each
COMPARE
4in Threaded Standoff Kit For 55 Gallon Drums
$47.50USD / Each
GoatThroat SO12 12in Threaded Standoff Kit For 55 Gallon Drums EW-75650-68 Mfr # SO12
COMPARE
12in Threaded Standoff Kit For 55 Gallon Drums
$62.00USD / Each
GoatThroat ST01 Spillproof Barrel Pump Siphon Tube, 95 cm; 1/Pk EW-75650-82 Mfr # ST01
COMPARE
Spillproof Barrel Pump Siphon Tube, 95 cm; 1/Pk
$43.00USD / Each
GoatThroat ST KIT Spillproof Barrel Pump Siphon Tube Kit with Connector; 1/Pk EW-75650-88 Mfr # ST KIT
COMPARE
Spillproof Barrel Pump Siphon Tube Kit with Connector; 1/Pk
$47.50USD / Each
GoatThroat 2M-BPR-T 57mm (2.25in) Buttress Adapter With O-ring EW-75650-90 Mfr # 2M-BPR-T
COMPARE
57mm (2.25in) Buttress Adapter With O-ring
$47.50USD / Each
GoatThroat 2M-BPR 63mm (2.5in) Buttress Adapter With O-ring EW-75650-92 Mfr # 2M-BPR
COMPARE
63mm (2.5in) Buttress Adapter With O-ring
$47.50USD / Each
GoatThroat 2M-BPR-E 70mm (2.75in) Buttress Adapter With O-ring EW-75650-94 Mfr # 2M-BPR-E
COMPARE
70mm (2.75in) Buttress Adapter With O-ring
$47.50USD / Each
Showing 1- 8 of 8