GoatThroat Spillproof Barrel Pumps
Pump wide selection of fluids from 5 gallon pails to 55 gallon barrels
  • Choice of four seal materials to handle over 800 liquids and chemicals
  • Guaranteed for 1 year. Life expectancy 7–10 years
  • Internal safety relief valve prevents overpressurization
  • Drums are Resource Conservation and Recovery Act (RCRA) ready
The GoatThroat pumps are designed to handle a wide range of liquids—wetted parts are 100% nonreactive, food-grade polypropylene. Choose nitrile seals for petroleum-based products including oil, diesel fuel, and glycerin. The EPDM seals are ideal for general-purpose chemicals including some acids, acetates, alcohols, and phosphates. Santoprene® seals are ideal for acetic acid, acetone, and hydrocyanic acid among others. Viton® seals were developed for more aggressive chemicals including bromine, sulfuric acid, ethylene, nitric acid, gasoline, and pesticides.
$259.50 - $365.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Wetted Materials
Availability Pricing
GoatThroat GT100-C/STANDOFF Pump With Nitrile Seal, 4in Standoff EW-75650-10 Mfr # GT100-C/STANDOFF
COMPARE
4.5 Body: Polypropylene; Seals: Nitrile
$259.50USD / Each
GoatThroat GT200-C/STANDOFF Pump With Epdm Seal, 4in Standoff EW-75650-12 Mfr # GT200-C/STANDOFF
COMPARE
4.5 Body: Polypropylene; Seals: EPDM
$265.00USD / Each
GoatThroat GT200S-C/STANDOFF Pump With Santoprene Seal, 4in; Standoff EW-75650-14 Mfr # GT200S-C/STANDOFF
COMPARE
4.5 Body: Polypropylene; Seals: Santoprene
$276.00USD / Each
GoatThroat GT300-C/STANDOFF Pump With Viton Seal, 4in; Standoff EW-75650-16 Mfr # GT300-C/STANDOFF
COMPARE
4.5 Body: Polypropylene; Seals: Viton
$365.00USD / Each